Assay Details
Gene Symbol: RP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203288.1 | NP_976033.1 | 1 - 2 | 164 | 165 | |
GenBank mRNA | BC025928.1 | - | 1 - 2 | 164 | 4903338 | 165 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001266.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07019751.hg.1 | TC07001266.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07019751.hg.1 | ENST00000297157.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07019751.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07019749.hg.1 | TC07001266.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07019749.hg.1 | ENST00000297157.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07019749.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07019752.hg.1 | TC07001266.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07019752.hg.1 | ENST00000297157.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07019752.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010697.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176188.hg.1 | TC0700010697.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000448915.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176185.hg.1 | TC0700010697.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000297157.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000474370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | ENST00000492391.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | NM_203288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700176187.hg.1 | TC0700010697.hg.1 | Clariom D TC |
