Assay Details
| Gene Symbol: LOC101927811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110119.1 | 2 - 3 | 203 | 111 | ||
| GenBank mRNA | AV707592.1 | - | 2 - 3 | 135 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927811| Gene Symbol: LOC101927811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110119.1 | 2 - 3 | 203 | 111 | ||
| GenBank mRNA | AV707592.1 | - | 2 - 3 | 135 | 111 | |
Set Membership: |
|