Assay Details
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045677.1 | 2 - 3 | 255 | 116 | ||
GenBank mRNA | AY122474.1 | - | 2 - 3 | 256 | 116 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | defensin beta 121 | DEFB121 |
DEFB21, ESC42RELC |
-2.4kb | Anti-sense | Chr. 20 - 31404845 - 31418522 on NCBI Build GRCh38 |
Downstream | defensin beta 123 | DEFB123 |
DEFB-23, DEFB23, ESC42-RELD |
11.9kb | Sense | Chr. 20 - 31440519 - 31450257 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000751.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31777305 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 447040 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 309916 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375626 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3417602 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3804 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3803 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18480 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26643 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P03505414 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P41303475 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375626 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3417602 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3375626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777305 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31777303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11130246 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03505414 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41303475 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P22378537 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03505414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130281 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17186348 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130284 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130254 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130232 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130245 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130242 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130261 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130226 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130292 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130230 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130279 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130224 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130268 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130263 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130257 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130239 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130235 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130266 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130286 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130237 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130228 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11130282 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17186347 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03505414 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1562167_PM_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR20010061.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010061.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010061.hg.1 | TC20000751.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010058.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010058.hg.1 | ENST00000545884.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010058.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010058.hg.1 | TC20000751.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20010060.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010060.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010060.hg.1 | TC20000751.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008809.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161514.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161514.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161514.hg.1 | TC2000008809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161514.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161514.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | TC2000008809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | ENST00000569013.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161510.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161513.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161513.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161513.hg.1 | TC2000008809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161513.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161513.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161509.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161509.hg.1 | NR_045677.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000161509.hg.1 | TC2000008809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000161509.hg.1 | ENST00000433453.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000161509.hg.1 | NR_045677.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1562167_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1562167_a_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1562167_a_at4 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018176 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A018176 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 698043 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2197228 | AY122474.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2197228 | AY122474.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2197228 | NR_002577.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2197228 | AY122474.1 | human_transcriptome |
