Assay Details
| Gene Symbol: LOC101928009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110685.1 | 1 - 2 | 121 | 80 | ||
| GenBank mRNA | BC038426.1 | - | 1 - 2 | 121 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small proline rich protein 2G | SPRR2G | -23.6kb | Anti-sense | Chr. 1 - 153149582 - 153195377 on NCBI Build GRCh38 | |
| Downstream | late cornified envelope like proline rich 1 | LELP1 | 26.8kb | Sense | Chr. 1 - 153203430 - 153205125 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928009
Genomic Map