Assay Details
| Gene Symbol: POU6F2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023033.1 | - | 2 - 3 | 294 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
VPS41, HOPS complex subunit |
VPS41 |
HVPS41, HVSP41, hVps41p |
-92.9kb | Sense | Chr. 7 - 38722974 - 38909200 on NCBI Build GRCh38 |
| Downstream | POU6F2 antisense RNA 1 | POU6F2-AS1 | 399.4kb | Sense | Chr. 7 - 39404598 - 39406346 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

POU6F2-AS2
Genomic Map