Assay Details
Gene Symbol: SAA3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026576.1 | 1 - 2 | 144 | 73 | ||
GenBank mRNA | AY209188.1 | - | 1 - 2 | 144 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001462.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 492846 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P556318 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P556318 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 32885 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31677 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P556318 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P556318 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670994 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671018 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070755 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671011 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670997 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671014 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070743 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671002 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070755 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671018 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671011 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670994 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07670996 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28070751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424685 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P556318 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19167581 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P556318 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19167581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16424685 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670966 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16424684 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670977 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671023 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670956 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670976 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670993 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670961 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670972 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671011 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670999 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671001 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670960 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670974 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671014 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670967 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670964 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670970 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670998 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671004 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670981 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671003 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670996 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671020 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671017 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671007 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671012 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671009 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671024 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670963 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07670973 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19167581 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7946974 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7946976 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7946975 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7946973 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7946972 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 222226_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11753559_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11753559_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11018471.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018471.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018471.hg.1 | TC11001462.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11018472.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018472.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018472.hg.1 | TC11001462.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11018473.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018473.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018473.hg.1 | TC11001462.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203849.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203849.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203849.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203849.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203849.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100203844.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203844.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203844.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203844.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203844.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100203841.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203841.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203841.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203841.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203841.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013151.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203848.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203842.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | ENST00000533267.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | ENST00000534768.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | NR_026576.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100203845.hg.1 | TC1100013151.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 222226_at | - | - |
Human Genome U133 Plus 2.0 Array | 222226_PM_at | - | - |
Human Genome U133A 2.0 Array | 222226_PM_at | - | - |
Human Genome U133A 2.0 Array | 222226_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw564301 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 222226_at7 | - | - |
Human Genome U133 Set | 222226_at | - | - |
Human Genome U133 Set | 222226_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044990 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044990 | - | - |
