Assay Details
| Gene Symbol: LINC01500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI520443.1 | - | 1 - 2 | 258 | 130 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000553762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000555378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | ENST00000556815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007323.hg.1 | NR_110547.1 | Refseq |

LINC01500
Genomic Map