Assay Details
| Gene Symbol: ANO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142649.1 | NP_001136121.1 | 6 - 7 | 675 | 154 | |
| NM_213599.2 | NP_998764.1 | 6 - 7 | 678 | 154 | ||
| XM_005252820.3 | XP_005252877.2 | 5 - 6 | 782 | 154 | ||
| XM_005252821.3 | XP_005252878.2 | 5 - 6 | 779 | 154 | ||
| XM_005252822.4 | XP_005252879.1 | 6 - 7 | 563 | 154 | ||
| XM_011519949.2 | XP_011518251.1 | 4 - 5 | 731 | 154 | ||
| GenBank mRNA | AB125267.1 | - | 6 - 7 | 678 | 154 | |
| AL832374.1 | - | 6 - 7 | 700 | 154 | ||
| AL833271.1 | - | 6 - 7 | 699 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000261.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11003510.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003510.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003510.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003510.hg.1 | TC11000261.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149839.hg.1 | TC1100007063.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000324559.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | ENST00000532043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_001142649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007063.hg.1 | NM_213599.2 | Refseq |

ANO5
Genomic Map