Assay Details
| Gene Symbol: LOC101927179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125881.1 | 1 - 2 | 161 | 138 | ||
| GenBank mRNA | AL599247.1 | - | 1 - 2 | 162 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927179| Gene Symbol: LOC101927179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125881.1 | 1 - 2 | 161 | 138 | ||
| GenBank mRNA | AL599247.1 | - | 1 - 2 | 162 | 138 | |
Set Membership: |
|