Assay Details
| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG215598.1 | - | 2 - 3 | 182 | 91 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 116822 | - | - |

MIR4500HG
Genomic Map