Assay Details
| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG215598.1 | - | 3 - 4 | 236 | 65 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000436290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000441617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000450888.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000453832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000606590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | ENST00000607309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009405.hg.1 | NR_033829.1 | Refseq |

MIR4500HG
Genomic Map