| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037188.hg.1 |
TC02002348.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037188.hg.1 |
ENST00000354183.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037188.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037188.hg.1 |
ENST00000409185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037188.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037186.hg.1 |
TC02002348.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037186.hg.1 |
ENST00000354183.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037186.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037186.hg.1 |
ENST00000409185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037186.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037187.hg.1 |
TC02002348.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037187.hg.1 |
ENST00000354183.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037187.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02037187.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204388.hg.1 |
TC0200014288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014288.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
ENST00000409185.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204385.hg.1 |
TC0200014288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200204387.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204387.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204387.hg.1 |
ENST00000389915.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200204387.hg.1 |
NM_001009993.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200204387.hg.1 |
TC0200014288.hg.1 |
Clariom D TC |