Assay Details
| Gene Symbol: LOC100507654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034289.1 | - | 2 - 3 | 128 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gamma-glutamylamine cyclotransferase | GGACT | A2LD1 | Sense | Chr. 13 - 100530164 - 100588792 on NCBI Build GRCh38 | |
| Downstream | transmembrane and tetratricopeptide repeat containing 4 | TMTC4 | 24.9kb | Sense | Chr. 13 - 100603836 - 100675025 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000376250.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000454752.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000455100.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000464500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000467518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000471912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000492399.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000608779.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | ENST00000629479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_001195087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009637.hg.1 | NM_033110.2 | Refseq |

LOC100507654
Genomic Map