Assay Details
| Gene Symbol: LINC01097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035722.1 | - | 2 - 3 | 215 | 89 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |

LINC01097
Genomic Map