Assay Details
| Gene Symbol: LINC01097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034054.1 | 3 - 4 | 549 | 64 | ||
| GenBank mRNA | AK096368.1 | - | 3 - 4 | 549 | 64 | |
| BC035722.1 | - | 3 - 4 | 374 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB28, member RAS oncogene family |
RAB28 | CORD18 | -42.0kb | Sense | Chr. 4 - 13293479 - 13484365 on NCBI Build GRCh38 |
| Downstream | NK3 homeobox 2 | NKX3-2 |
BAPX1, NKX3.2, NKX3B, SMMD |
12.6kb | Sense | Chr. 4 - 13540830 - 13544490 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001044.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088511 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088528 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088549 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15547065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02936196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23088522 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 142987 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 320271 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 887922 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3637366 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35553 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P14605726 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3637366 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3637366 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00922434 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00922430 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P14605726 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P704696 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562683_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014290.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014290.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014290.hg.1 | ENST00000536697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014290.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014289.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014289.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014289.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014293.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014293.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014293.hg.1 | ENST00000536697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014293.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014296.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014296.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014296.hg.1 | ENST00000536697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014296.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014291.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014291.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014291.hg.1 | ENST00000536697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014291.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014297.hg.1 | TC04001044.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014297.hg.1 | ENST00000505835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014297.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169078.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169082.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169075.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169081.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169073.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169073.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169073.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169073.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169073.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000627575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | ENST00000628853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169076.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400169083.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169083.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169083.hg.1 | ENST00000627163.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400169083.hg.1 | NR_034054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400169083.hg.1 | TC0400010082.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1562683_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562683_PM_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562683_a_at1 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023963 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023963 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 231097 | - | - |

LINC01097
Genomic Map