| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003212.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003212.hg.1 |
ENST00000544540.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003212.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10003212.hg.1 |
TC10000287.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000150644.hg.1 |
TC1000007458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000389583.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000460364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
ENST00000544540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007458.hg.1 |
NM_001123376.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010478.hg.1 |
ENST00000436877.2 |
Ensembl |