Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139444.2 | NP_001132916.1 | 2 - 3 | 241 | 112 | |
| GenBank mRNA | BX280366.1 | - | 2 - 3 | 241 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002049.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06025586.hg.1 | TC06002049.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025586.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025586.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025581.hg.1 | TC06002049.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025581.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025581.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025581.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025585.hg.1 | TC06002049.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025585.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025585.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025587.hg.1 | TC06002049.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025587.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06025587.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189483.hg.1 | TC0600012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600189489.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189489.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189489.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189489.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189489.hg.1 | TC0600012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600189490.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189490.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189490.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189490.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189490.hg.1 | TC0600012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189488.hg.1 | TC0600012968.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600189487.hg.1 | TC0600012968.hg.1 | Clariom D TC |

TRAPPC3L
Genomic Map