Assay Details
| Gene Symbol: CLVS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010852.3 | NP_001010852.2 | 2 - 3 | 1724 | 92 | |
| XM_017010266.1 | XP_016865755.1 | 2 - 3 | 3426 | 92 | ||
| GenBank mRNA | AK095527.1 | - | 2 - 3 | 1728 | 92 | |
| AK295158.1 | - | 2 - 3 | 186 | 92 | ||
| BC101376.1 | - | 2 - 3 | 1258 | 40025886 | 92 | |
| BC101377.2 | - | 2 - 3 | 1260 | 40025889 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000951.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK295158.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06011086.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011086.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011086.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011086.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011083.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011083.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011083.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011083.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011084.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011084.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011084.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011084.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011082.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011082.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011082.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011080.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011080.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011080.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011081.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011081.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011081.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011081.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011085.hg.1 | TC06000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011085.hg.1 | ENST00000275162.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011085.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011085.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163944.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163944.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163944.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163944.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163944.hg.1 | TC0600009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009336.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163942.hg.1 | TC0600009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163941.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163941.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163941.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163941.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163941.hg.1 | TC0600009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | ENST00000368438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163945.hg.1 | TC0600009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163939.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163939.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163939.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163939.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163939.hg.1 | TC0600009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163943.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163943.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163943.hg.1 | ENST00000275162.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163943.hg.1 | NM_001010852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163943.hg.1 | TC0600009336.hg.1 | Clariom D TC |

CLVS2
Genomic Map