Assay Details
| Gene Symbol: SLC6A19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003841.2 | NP_001003841.1 | 3 - 4 | 536 | 146 | |
| GenBank mRNA | AK290811.1 | - | 3 - 4 | 536 | 146 | |
| AY591756.1 | - | 3 - 4 | 480 | 146 | ||
| AY596807.1 | - | 3 - 4 | 480 | 146 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000016.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05000323.hg.1 | TC05000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000323.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000323.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000324.hg.1 | TC05000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000324.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000324.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144665.hg.1 | TC0500006493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144664.hg.1 | TC0500006493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000304460.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | ENST00000515652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006493.hg.1 | NM_001003841.2 | Refseq |

SLC6A19
Genomic Map