Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002042.1 | - | 1 - 2 | 152 | 156 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000356128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000368602.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | ENST00000437098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012968.hg.1 | NM_001139444.2 | Refseq |

TRAPPC3L
Genomic Map