Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104462.1 | 2 - 3 | 725 | 96 | ||
| GenBank mRNA | AL832235.1 | - | 2 - 3 | 725 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

CENPW
Genomic Map