Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286524.1 | NP_001273453.1 | 1 - 2 | 79 | ||
| GenBank mRNA | AI207187.1 | - | 2 - 3 | 260 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368326.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | ENST00000368328.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001012507.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NM_001286525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009381.hg.1 | NR_104462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164379.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164379.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164379.hg.1 | ENST00000368325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164379.hg.1 | NM_001286524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164379.hg.1 | TC0600009381.hg.1 | Clariom D TC |

CENPW
Genomic Map