Assay Details
| Gene Symbol: LOC101928231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125852.1 | 1 - 2 | 689 | 62 | ||
| GenBank mRNA | BC040308.1 | - | 1 - 2 | 691 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928231
Genomic Map