Assay Details
| Gene Symbol: LOC101928231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125852.1 | 3 - 4 | 927 | 72 | ||
| GenBank mRNA | BC040308.1 | - | 3 - 4 | 929 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928231| Gene Symbol: LOC101928231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125852.1 | 3 - 4 | 927 | 72 | ||
| GenBank mRNA | BC040308.1 | - | 3 - 4 | 929 | 72 | |
Set Membership: |
|