Assay Details
| Gene Symbol: LOC101929261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134277.1 | 1 - 2 | 105 | 85 | ||
| GenBank mRNA | BC030121.2 | - | 1 - 2 | 105 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | golgin A6 family-like 1 pseudogene | LOC442132 | -40.3kb | Sense | Chr. 5 - 7301330 - 7306714 on NCBI Build GRCh38 | |
| Downstream | adenylate cyclase 2 | ADCY2 |
AC2, HBAC2 |
48.3kb | Anti-sense | Chr. 5 - 7396230 - 7830081 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929261
Genomic Map