Assay Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090937.1 | - | 2 - 3 | 739 | 80 | |
BX640850.1 | - | 2 - 3 | 575 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000391411.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000412778.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000426261.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000468381.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000535846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000536958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000538604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000540508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | ENST00000582941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010101.hg.1 | NM_001113434.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007274.hg.1 | ENST00000579239.1 | Ensembl |
