Assay Details
| Gene Symbol: LOC101928516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110856.1 | 3 - 4 | 275 | 64 | ||
| GenBank mRNA | AF086303.1 | - | 3 - 4 | 275 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928516| Gene Symbol: LOC101928516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110856.1 | 3 - 4 | 275 | 64 | ||
| GenBank mRNA | AF086303.1 | - | 3 - 4 | 275 | 64 | |
Set Membership: |
|