Assay Details
| Gene Symbol: FAM102B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010883.2 | NP_001010883.2 | 9 - 10 | 1051 | 100 | |
| GenBank mRNA | BC127898.1 | - | 9 - 10 | 1051 | 40132918 | 100 |
| BC140863.1 | - | 9 - 10 | 907 | 9021731 | 100 | |
| BC140865.1 | - | 9 - 10 | 907 | 9021733 | 100 | |
| BC150530.1 | - | 9 - 10 | 1051 | 40132919 | 100 | |
| CR749397.1 | - | 6 - 7 | 622 | 100 | ||
| CR936815.1 | - | 2 - 3 | 4035 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000931.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014615.hg.1 | TC01000931.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014615.hg.1 | ENST00000370035.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014615.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014615.hg.1 | ENST00000437902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014615.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009324.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000370035.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000405454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | ENST00000483371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | NM_001010883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166402.hg.1 | TC0100009324.hg.1 | Clariom D TC |

FAM102B
Genomic Map