Assay Details
| Gene Symbol: SYCE1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129979.1 | NP_001123451.1 | 9 - 10 | 579 | 117 | |
| GenBank mRNA | BC035731.1 | - | 1 - 2 | 93 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16002060.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008896.hg.1 | TC16002060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008896.hg.1 | ENST00000378644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008896.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008895.hg.1 | TC16002060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008895.hg.1 | ENST00000378644.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008895.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011433.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000563157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | ENST00000568925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191563.hg.1 | TC1600011433.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191560.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191560.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191560.hg.1 | ENST00000378644.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191560.hg.1 | NM_001129979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191560.hg.1 | TC1600011433.hg.1 | Clariom D TC |

SYCE1L
Genomic Map