Assay Details
| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080534.1 | NP_001074003.1 | 14 - 15 | 4487 | 125 | |
| XM_005254394.4 | XP_005254451.1 | 15 - 16 | 5438 | 125 | ||
| XM_017022220.1 | XP_016877709.1 | 19 - 20 | 5358 | 125 | ||
| XM_017022221.1 | XP_016877710.1 | 18 - 19 | 5238 | 125 | ||
| XM_017022222.1 | XP_016877711.1 | 17 - 18 | 5136 | 125 | ||
| XM_017022223.1 | XP_016877712.1 | 15 - 16 | 10664 | 125 | ||
| XM_017022224.1 | XP_016877713.1 | 14 - 15 | 5364 | 125 | ||
| XM_017022225.1 | XP_016877714.1 | 12 - 13 | 2789 | 125 | ||
| GenBank mRNA | AK091491.1 | - | 11 - 12 | 1463 | 125 | |
| AK293996.1 | - | 4 - 5 | 404 | 125 | ||
| BC039494.1 | - | 7 - 8 | 1062 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000429.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003938.hg.1 | ENST00000260323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003938.hg.1 | ENST00000537900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003938.hg.1 | ENST00000545554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003938.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003938.hg.1 | TC15000429.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000539562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000559093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000560537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007294.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | ENST00000260323.15 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | ENST00000561210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | NM_001080534.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151955.hg.1 | TC1500007294.hg.1 | Clariom D TC |

UNC13C
Genomic Map