Assay Details
| Gene Symbol: LOC101929076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120345.1 | 1 - 2 | 168 | 69 | ||
| GenBank mRNA | BC033162.1 | - | 1 - 2 | 168 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SKI family transcriptional corepressor 1 | SKOR1 |
CORL1, FUSSEL15, LBXCOR1 |
-2.5kb | Anti-sense | Chr. 15 - 67819704 - 67834561 on NCBI Build GRCh38 |
| Downstream |
RNA, U6 small nuclear 1 |
RNU6-1 |
RNU6, RNU6A, U6, U6-1 |
1.1kb | Sense | Chr. 15 - 67839940 - 67840045 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929076
Genomic Map