Assay Details
| Gene Symbol: LINC00284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026955.1 | 2 - 3 | 409 | 62 | ||
| GenBank mRNA | AK001938.1 | - | 1 - 2 | 1579 | 62 | |
| AK001941.1 | - | 2 - 3 | 256 | 62 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000423211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000587571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000591799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000592085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000609808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000613822.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000616953.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | ENST00000617476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009958.hg.1 | NR_026955.1 | Refseq |

LINC00284
Genomic Map