Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004318.2 | NP_001004318.2 | 5 - 6 | 899 | 64 | |
| XM_011526966.2 | XP_011525268.1 | 5 - 6 | 951 | 64 | ||
| XM_011526967.2 | XP_011525269.1 | 5 - 6 | 899 | 64 | ||
| XM_011526969.1 | XP_011525271.1 | 5 - 6 | 899 | 64 | ||
| XM_017026807.1 | XP_016882296.1 | 5 - 6 | 1750 | 64 | ||
| GenBank mRNA | AK131245.1 | - | 5 - 6 | 899 | 64 | |
| BC127841.1 | - | 8 - 9 | 64 | |||
| BC136721.1 | - | 4 - 5 | 701 | 9020711 | 64 | |
| BC136722.1 | - | 4 - 5 | 701 | 9020712 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000536.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19007284.hg.1 | ENST00000331256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19007284.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19007284.hg.1 | TC19000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | TC1900008041.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900160430.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000331256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000594229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | ENST00000601575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008041.hg.1 | NM_001004318.2 | Refseq |

ACP7
Genomic Map