Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004318.2 | NP_001004318.2 | 6 - 7 | 970 | 90 | |
| XM_011526966.2 | XP_011525268.1 | 6 - 7 | 1022 | 90 | ||
| XM_011526967.2 | XP_011525269.1 | 6 - 7 | 970 | 90 | ||
| XM_011526969.1 | XP_011525271.1 | 6 - 7 | 970 | 90 | ||
| XM_017026807.1 | XP_016882296.1 | 6 - 7 | 1821 | 90 | ||
| XM_017026808.1 | XP_016882297.1 | 5 - 6 | 883 | 90 | ||
| GenBank mRNA | AK131245.1 | - | 6 - 7 | 970 | 90 | |
| BC127841.1 | - | 7 - 8 | 90 | |||
| BC136721.1 | - | 5 - 6 | 772 | 9020711 | 90 | |
| BC136722.1 | - | 5 - 6 | 772 | 9020712 | 90 | |
| BC150149.1 | - | 6 - 7 | 90 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000536.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017026808.1 transcripts.
Set Membership: |
|

ACP7
Genomic Map