Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315530.1 | NP_001302459.1 | 1 - 2 | 387 | 105 | |
| GenBank mRNA | BI603704.1 | - | 1 - 2 | 391 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315530.1 transcripts.
Set Membership: |
|

RPL26
Genomic Map