| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003989.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18003989.hg.1 |
TC18000379.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155743.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155743.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155743.hg.1 |
TC1800008151.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800155743.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155743.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
ENST00000588672.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008151.hg.1 |
NR_040031.1 |
Refseq |