Assay Details
| Gene Symbol: TIMM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001563.2 | NP_001001563.1 | 3 - 4 | 729 | 71 | |
| XM_011527491.2 | XP_011525793.1 | 3 - 4 | 272 | 71 | ||
| GenBank mRNA | AY551341.1 | - | 3 - 4 | 287 | 71 | |
| BC009072.1 | - | 3 - 4 | 261 | 71 | ||
| BC010736.1 | - | 3 - 4 | 282 | 71 | ||
| BC050082.1 | - | 3 - 4 | 305 | 71 | ||
| BC121147.1 | - | 3 - 4 | 657 | 40121258 | 71 | |
| BQ925812.1 | - | 3 - 4 | 316 | 71 | ||
More Information
Set Membership: |
|

TIMM50
Genomic Map