Assay Details
| Gene Symbol: RAPGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263358.3 | XP_005263415.1 | 2 - 3 | 235 | 79 | |
| XM_005263359.3 | XP_005263416.1 | 2 - 3 | 237 | 79 | ||
| XM_005263360.3 | XP_005263417.1 | 2 - 3 | 229 | 79 | ||
| XM_005263361.3 | XP_005263418.1 | 2 - 3 | 235 | 79 | ||
| XM_006714420.3 | XP_006714483.1 | 2 - 3 | 235 | 79 | ||
| XM_006714421.3 | XP_006714484.1 | 2 - 3 | 202 | 79 | ||
| XM_017008856.1 | XP_016864345.1 | 2 - 3 | 236 | 79 | ||
More Information
Set Membership: |
|

RAPGEF2
Genomic Map