Assay Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109977.1 | NP_001103447.1 | 6 - 7 | 1473 | 101 | |
| XM_005263199.1 | XP_005263256.1 | 5 - 6 | 1348 | 101 | ||
| XM_011532220.1 | XP_011530522.1 | 4 - 5 | 1226 | 101 | ||
| XM_011532221.2 | XP_011530523.1 | 6 - 7 | 1546 | 101 | ||
| XM_011532222.2 | XP_011530524.1 | 5 - 6 | 1468 | 101 | ||
| XM_011532223.2 | XP_011530525.1 | 4 - 5 | 1343 | 101 | ||
| XM_011532224.2 | XP_011530526.1 | 4 - 5 | 1042 | 101 | ||
| GenBank mRNA | BC014937.1 | - | 4 - 5 | 1057 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000754.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04010461.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010461.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010461.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010461.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010461.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010463.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010463.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010463.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010463.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010467.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010467.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010467.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010467.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010469.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010469.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010469.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010469.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010466.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010466.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010466.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010466.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010468.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010468.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010468.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010468.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010456.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010456.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010456.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010456.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010457.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010457.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010457.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010457.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010462.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010462.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010462.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010462.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010452.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010452.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010452.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010452.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010453.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010453.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010453.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010453.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010455.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010455.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010455.hg.1 | ENST00000508198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010455.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161734.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161737.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161752.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161735.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161738.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161745.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161750.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161739.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161743.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161746.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161749.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161751.hg.1 | TC0400009001.hg.1 | Clariom D TC |

FAM160A1
Genomic Map