Assay Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109977.1 | NP_001103447.1 | 8 - 9 | 1719 | 149 | |
| XM_005263199.1 | XP_005263256.1 | 7 - 8 | 1594 | 149 | ||
| XM_011532220.1 | XP_011530522.1 | 6 - 7 | 1472 | 149 | ||
| XM_011532221.2 | XP_011530523.1 | 8 - 9 | 1792 | 149 | ||
| XM_011532222.2 | XP_011530524.1 | 7 - 8 | 1714 | 149 | ||
| XM_011532223.2 | XP_011530525.1 | 6 - 7 | 1589 | 149 | ||
| XM_011532224.2 | XP_011530526.1 | 6 - 7 | 1288 | 149 | ||
| GenBank mRNA | BC014937.1 | - | 6 - 7 | 1303 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000754.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010471.hg.1 | TC04000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010471.hg.1 | ENST00000435205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010471.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010471.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161754.hg.1 | TC0400009001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |

FAM160A1
Genomic Map