Assay Details
| Gene Symbol: LINC01447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108090.1 | 1 - 2 | 394 | 72 | ||
| GenBank mRNA | AK056440.1 | - | 1 - 2 | 394 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01447| Gene Symbol: LINC01447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108090.1 | 1 - 2 | 394 | 72 | ||
| GenBank mRNA | AK056440.1 | - | 1 - 2 | 394 | 72 | |
Set Membership: |
|