Assay Details
| Gene Symbol: LINC00639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125018.1 | - | 4 - 5 | 3985 | 88 | |
| AK127318.1 | - | 2 - 3 | 3085 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-type lectin domain family 14 member A | CLEC14A |
C14orf27, CEG1, EGFR-5 |
-506.6kb | Sense | Chr. 14 - 38254000 - 38256370 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105370457 | LOC105370457 | 43.3kb | Anti-sense | Chr. 14 - 38878730 - 38919201 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000553932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000554732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000556116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | ENST00000557440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008998.hg.1 | NR_039982.1 | Refseq |

LINC00639
Genomic Map