Assay Details
| Gene Symbol: NLRP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319967.1 | NP_001306896.1 | 1 | 1981 | 103 | |
| GenBank mRNA | AK026387.1 | - | 2 | 1480 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NLRP2B
Genomic Map