Assay Details
| Gene Symbol: LOC441239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516745.2 | XP_011515047.1 | 1 - 2 | 654 | 130 | |
| GenBank mRNA | BC063653.1 | - | 1 - 2 | 654 | 130 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation elongation factor 1 delta pseudogene 4 | EEF1DP4 | -24.6kb | Anti-sense | Chr. 7 - 64862951 - 64864450 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 117 | ZNF117 |
H-plk, HPF9 |
84.4kb | Sense | Chr. 7 - 64974452 - 64991036 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC441239
Genomic Map