Assay Details
| Gene Symbol: ENO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006717835.3 | XP_006717898.1 | 13 - 14 | 3065 | 79 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |

ENO4
Genomic Map