Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198992.3 | NP_945343.1 | 1 - 2 | 443 | 104 | |
| GenBank mRNA | AY198413.1 | - | 1 - 2 | 444 | 104 | |
| BC101024.2 | - | 1 - 2 | 270 | 40007421 | 104 | |
| BC101025.1 | - | 1 - 2 | 270 | 40007422 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001380.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC101025.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
chemical synaptic transmission
sensory perception of smell
synaptic vesicle exocytosis
regulation of calcium ion-dependent exocytosis
positive regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
protein homodimerization activity
protein heterodimerization activity
vesicle fusion
chemical synaptic transmission
sensory perception of smell
synaptic vesicle exocytosis
regulation of calcium ion-dependent exocytosis
positive regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12017951.hg.1 | ENST00000228567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017951.hg.1 | ENST00000535526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017951.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017951.hg.1 | TC12001380.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017947.hg.1 | ENST00000228567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017947.hg.1 | ENST00000535526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017947.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017947.hg.1 | TC12001380.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017952.hg.1 | ENST00000228567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017952.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017952.hg.1 | TC12001380.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017950.hg.1 | ENST00000228567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017950.hg.1 | ENST00000535526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017950.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017950.hg.1 | TC12001380.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178289.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010352.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178284.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178290.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200178286.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178286.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178286.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178286.hg.1 | ENST00000567656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178286.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200178292.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178292.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178292.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178292.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178292.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200178287.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178287.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178287.hg.1 | TC1200010352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | ENST00000228567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | ENST00000539102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | NM_198992.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200178291.hg.1 | TC1200010352.hg.1 | Clariom D TC |

SYT10
Genomic Map