| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000567656.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
ENST00000567656.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017945.hg.1 |
ENST00000228567.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017945.hg.1 |
ENST00000535526.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017945.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017945.hg.1 |
TC12001380.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017946.hg.1 |
ENST00000228567.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017946.hg.1 |
ENST00000535526.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017946.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12017946.hg.1 |
TC12001380.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200178282.hg.1 |
TC1200010352.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000567656.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
ENST00000567656.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010352.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
ENST00000228567.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
ENST00000539102.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
NM_198992.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200178283.hg.1 |
TC1200010352.hg.1 |
Clariom D TC |