Assay Details
| Gene Symbol: TMEM30C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028357.1 | 2 - 3 | 164 | 130 | ||
| GenBank mRNA | AB249666.1 | - | 2 - 3 | 164 | 130 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000505.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447511 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15468784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585688 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22585690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02447526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15468785 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 246826 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 460565 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3225357 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37887 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16333677 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3225357 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3225357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585645 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447444 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447441 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447476 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585645 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447441 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22585648 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447444 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02447476 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 158035 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 4596 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16333662 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36376865 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P14309192 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16333677 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16333677 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03009477.hg.1 | TC03000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009477.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009477.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000476804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008140.hg.1 | NR_028358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157964.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157964.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157964.hg.1 | ENST00000489487.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157964.hg.1 | NR_028357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157964.hg.1 | TC0300008140.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022922 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022923 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022923 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022922 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693655 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1693655 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693655 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693655 | - | - |

TMEM30C
Genomic Map