Assay Details
| Gene Symbol: DNAH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291661.1 | NP_001278590.1 | 39 - 40 | 6214 | 93 | |
| XM_011533466.1 | XP_011531768.1 | 39 - 40 | 6199 | 93 | ||
| XM_011533467.1 | XP_011531769.1 | 39 - 40 | 6166 | 93 | ||
| XM_011533468.1 | XP_011531770.1 | 39 - 40 | 6199 | 93 | ||
| XM_011533469.1 | XP_011531771.1 | 39 - 40 | 6199 | 93 | ||
| XM_011533471.2 | XP_011531773.1 | 39 - 40 | 6199 | 93 | ||
| XM_017005860.1 | XP_016861349.1 | 39 - 40 | 6199 | 93 | ||
| XM_017005861.1 | XP_016861350.1 | 38 - 39 | 6882 | 93 | ||
| XM_017005862.1 | XP_016861351.1 | 18 - 19 | 2774 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

DNAH12
Genomic Map