Assay Details
| Gene Symbol: GADL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207359.2 | NP_997242.2 | 4 - 5 | 465 | 92 | |
| XM_017006297.1 | XP_016861786.1 | 4 - 5 | 863 | 92 | ||
| GenBank mRNA | AL832766.1 | - | 4 - 5 | 465 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001268.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03021634.hg.1 | TC03001268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03021634.hg.1 | ENST00000282538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03021634.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03021634.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300177334.hg.1 | TC0300010620.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000282538.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000454381.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | ENST00000498387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010620.hg.1 | NM_207359.2 | Refseq |

GADL1
Genomic Map